DL-Ornithine hydrochloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAO0063-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:168.62 g/mol
Merck Index:14,06874
EINECS:213-956-0
CAS Number:1069-31-4
Formula:C₅H₁₂N₂O₂·HCl
MDL Number:MFCD00065398