Pyridoxal hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:65-22-5
:
Formula:C₈H₁₀ClNO₃
MDL Number:MFCD00012809
:203.62 g/mol
Merck Index:14,07978
EINECS:200-602-5
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Water:very faint turbidity
Specification Test Results
Frequently Bought Together