

cis-1,2,3,6-Tetrahydrophthalic anhydride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:935-79-5
:
Formula:C₈H₈O₃
MDL Number:MFCD00005916
:152.15 g/mol
EINECS:213-308-7
UN:2698
ADR:8,III
:GHS05
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Methanolysis Method):min. 98.0 %
- Melting point:101.0 to 104.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together