Cesium chloride ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7647-17-8
:
Formula:CsCl
MDL Number:MFCD00010955
:168.36 g/mol
Merck Index:14,02011
Boiling Pt:1303 °C (1013 hPa)
Melting Pt:646 °C
Density:3,7808 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-600-2
Specification Test Results
- Appearance:White powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
Specification Test Results
Related Information
Frequently Bought Together