Order Entry
Belgium
ContactUsLinkComponent
 
Yttrium(III) trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
 :  TCI
 
undefined
Yttrium(III) trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 % (calcd.on anh.substance)
  • Water:
    max. 10.0 %

 

Frequently Bought Together