1,2,4-Trimethylbenzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Pseudocumene
CAS Number:95-63-6
:
Formula:C₉H₁₂
MDL Number:MFCD00008527
:120.19 g/mol
Merck Index:14,07915
Boiling Pt:169°C
Melting Pt:–44 °C
Density:0,88 g/cm³ (20 °C)
Flash Pt:44°C
Storage Temperature:Ambient
EINECS:202-436-9
UN:3295
ADR:3,III
:GHS02
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together