Order Entry
Belgium
ContactUsLinkComponent
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Flammable
Health
Irritant
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Catalog # 84708.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Catalog # 84708.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Purity (GC):
    min. 99.9%
  • Acidity:
    max. 0.0001 meq/g
  • Alkalinity:
    max. 0.0002 meq/g
  • Evaporation residue:
    max. 0.0002%
  • Water:
    max. 0.01%
  • Color:
    max. 10 Hz
  • Fluorescence (as quinine at 365 nm):
    max. 1.0 ppb
  • Transmission at 285 nm:
    min. 15%
  • Transmission at 290 nm:
    min. 60%
  • Transmission at 300 nm:
    min. 80%
  • Transmission at 310 nm:
    min. 90%
  • Transmission at 335 nm:
    min. 96%
  • Transmission from 350 nm:
    min. 98%
  • Absorbance at 285 nm:
    max. 0.82
  • Absorbance at 290 nm:
    max. 0.22
  • Absorbance at 300 nm:
    max. 0.10
  • Absorbance at 310 nm:
    max. 0.05
  • Absorbance at 335 nm:
    max. 0.02
  • Absorbance from 350 nm:
    max. 0.01

Specification Test Results

About this item

 :
 :92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119471310-51
 :GHS02
 :GHS08
 :GHS07
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512