2-Chloro-1-fluoro-4-nitrobenzene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 3-Chloro-4-fluoronitrobenzene, 1-Chloro-2-fluoro-5-nitrobenzene
CAS Number:350-30-1
:
Formula:C₆H₃ClFNO₂
MDL Number:MFCD00007206
:175.55 g/mol
Boiling Pt:105°C
Melting Pt:41.5 °C
EINECS:206-499-3
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:41.0 to 45.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together