
3-Hydroxy-p-anisic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:645-08-9
:
Formula:C₈H₈O₄
MDL Number:MFCD00002507
:168.15 g/mol
Boiling Pt:327 °C (1013 hPa)
Melting Pt:258…259 °C
EINECS:211-430-5
:GHS07
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:252.0 to 256.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together