4-Nitrobenzoic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:62-23-7
:
Formula:C₇H₅NO₄
MDL Number:MFCD00007352
:167.12 g/mol
Merck Index:12,06686
Boiling Pt:359 °C (1013 hPa)
Melting Pt:121 °C
Density:1,65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-526-2
:GHS07
:GHS08
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Solubility in hot Methanol:almost transparency
- Melting point:235.0 to 245.0 deg-C
Specification Test Results
Frequently Bought Together