
4-Aminobenzoic acid ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:150-13-0
:
Formula:C₇H₇NO₂
MDL Number:MFCD00007894
:137.14 g/mol
Merck Index:14,00423
Boiling Pt:340 °C (1013 hPa)
Melting Pt:188 °C
Density:1,45 g/cm³ (20 °C)
EINECS:205-753-0
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
Related Information
Frequently Bought Together