2-Pentyne ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:627-21-4
:
Formula:C₅H₈
MDL Number:MFCD00009380
Boiling Pt:56°C
Molecular Weight:68.12 g/mol
Melting Pt:-109
Flash Pt:–20°C
EINECS:210-989-2
UN:3295
ADR:3 II
:GHS02
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together