2-Methyl-2-butene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:513-35-9
:26760-64-5
Formula:C₅H₁₀
MDL Number:MFCD00009276
:70.13 g/mol
Merck Index:14,00607
Boiling Pt:38°C
Melting Pt:–134°C
Density:0,6623 g/cm³ (20 °C)
Flash Pt:–45°C
Storage Temperature:Refrigerator
EINECS:208-156-3
UN:2460
ADR:3,II
:GHS02
:GHS07
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together