DL-α-Tocopherol (Vitamin E) ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:10191-41-0
:1406-18-4
Formula:C₂₉H₅₀O₂
MDL Number:MFCD00072051
Merck Index:14,09495
Flash Pt:240°C
:430.71 g/mol
EINECS:233-466-0
Specification Test Results
- Appearance:Yellow to Amber to Dark purple clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together