Order Entry
Belgium
ContactUsLinkComponent
 
n-Hexadecane ≥98.0% (by GC)
 :  TCI
 
undefined
n-Hexadecane ≥98.0% (by GC)
 :  TCI
 

 

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

Frequently Bought Together