Order Entry
Belgium
ContactUsLinkComponent
n-Heptane ≥99.3%, SPECTRONORM® for spectroscopy
n-Heptane ≥99.3%, SPECTRONORM® for spectroscopy
Supplier:  VWR Chemicals
Certificates
undefined
n-Heptane ≥99.3%, SPECTRONORM® for spectroscopy
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (GC):
    ≥ 99,3 %
  • Acidity:
    ≤ 0,0001 meq/g
  • Alkalinity:
    ≤ 0,0002 meq/g
  • Colouration:
    ≤ 10 APHA
  • Evaporation residue:
    ≤ 2 ppm
  • Water:
    ≤ 50 ppm
  • Absorbance (200 nm):
    ≤ 0,70
  • Absorbance (210 nm):
    ≤ 0,26
  • Absorbance (220 nm):
    ≤ 0,10
  • Absorbance (228 nm):
    ≤ 0,05
  • Absorbance (from 245 nm):
    ≤ 0,01
  • Transmittance (200 nm):
    ≥ 20 %
  • Transmittance (210 nm):
    ≥ 55 %
  • Transmittance (220 nm):
    ≥ 80 %
  • Transmittance (228 nm):
    ≥ 90 %
  • Transmittance (from 245 nm):
    ≥ 98 %
  • Fluorescence (as quinine) (254 nm):
    ≤ 1,0 ppb
  • Fluorescence (as quinine) (365 nm):
    ≤ 1,0 ppb

Specification Test Results

Frequently Bought Together