Order Entry
Belgium
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) ≥85.0% (by titrimetric analysis)
Irritant
Health
Sodium dodecyl sulphate (SDS) ≥85.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Health
Sodium dodecyl sulphate (SDS) ≥85.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 50.0 %
  • Purity (Neutralization back titration):
    min. 85.0 %
  • Average molecular weight:
    285 to 310

Specification Test Results

Frequently Bought Together