

2-Propanol ≥99.5% (by GC)
: TCI
:67-63-0
:
:(CH₃)₂CHOH
:MFCD00011674
:60.1 g/mol
:14,05208
:82°C
:–89 °C
:0,786 g/cm³ (20 °C)
:15°C
:Ambient
:200-661-7
:1219
:3,II
:01-2119457558-25
:GHS02
:GHS07
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.5 %
Frequently Bought Together