Order Entry
Belgium
ContactUsLinkComponent
1-Butanol ≥99.9%, SPECTRONORM® for spectroscopy
 
 
 
1-Butanol ≥99.9%, SPECTRONORM® for spectroscopy
  84709.290
   Safer Human and Environmental Health
 :  
undefined
 
 
 
1-Butanol ≥99.9%, SPECTRONORM® for spectroscopy
  84709.290
   Safer Human and Environmental Health
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Purity (GC):
    min. 99.9%
  • Water:
    max. 0.03%
  • Evaporation residue:
    max. 0.0002%
  • Fluorescence (as quinine at 254 nm):
    max. 1.0 ppb
  • Fluorescence (as quinine at 365 nm):
    max. 1.0 ppb
  • Transmittance at 210 nm:
    min. 25%
  • Transmittance at 220 nm:
    min. 60%
  • Transmittance at 230 nm:
    min. 70%
  • Transmittance at 240 nm:
    min. 85%
  • Transmittance at 245 nm:
    min. 90%
  • Transmittance at 270 nm:
    min. 98%

 

 

 :
 :74.12 g/mol
 :13,01539
 :117.6 °C (1013 hPa)
 :–89 °C
 :0.812 g/cm³ (20 °C)
 :30 °C
 :Ambient
 :200-751-6
 :1120
 :3,III
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
 :01-2119484630-38
 :GHS02
 :GHS07
 :GHS05
 :71-36-3
 :H₃C(CH₂)₂CH₂OH
 :MFCD00002964