1-Butanol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:71-36-3
:
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964
:74.12 g/mol
Merck Index:14,01540
Boiling Pt:118°C
Melting Pt:–89 °C
Density:0,812 g/cm³ (20 °C)
Flash Pt:37°C
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
REACH:01-2119484630-38
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together