Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Dichloromethane ≥99.8%, SupraSolv® for GC-MS
 :  Merck
 
undefined
 
 
Dichloromethane ≥99.8%, SupraSolv® for GC-MS
 :  Merck
 
 
 

 

  • Purity (GC):
    =99.8%
  • Identity(IR):
    Conforms
  • Evaporation Residue:
    = 5.0 mg/l
  • Water:
    = 0.01%
  • Color:
    =10 Hazen
  • GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)):
    = 3ng/ml
  • Suitability:
    Residue Analysis

 

Frequently Bought Together