Order Entry
Belgium
ContactUsLinkComponent
 
Irritant
Ammonium acetate for HPLC, J.T.Baker®
Catalog # 0599-08
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Ammonium acetate for HPLC, J.T.Baker®
Catalog # 0599-08
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 97 %
  • Assay (CH₃COONH₄) (by acid-base titration):
    ≥97 %
  • Insoluble Matter:
    ≤0.005 %
  • Residue after Ignition:
    ≤0.010 %
  • pH of 5% Solution at 25°C:
    6.7 - 7.3
  • Nitrate (NO₃):
    ≤0.001 %
  • Sulfate (SO₄):
    ≤0.001 %
  • UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 350 nm:
    ≤0.010
  • UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 280 nm:
    ≤0.010
  • UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 254 nm:
    ≤0.020
  • Chloride (Cl):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

About this item

 :
 :77.08 g/mol
Merck Index:13,00495
Melting Pt:114 °C
Density:1.17 g/cm³ (20 °C)
Flash Pt:136 °C
EINECS:211-162-9
 :GHS07
Storage Temperature:Ambient
CAS Number:631-61-8
Formula:H₃CCOONH₄
MDL Number:MFCD00013066