Acetone ≥99.9%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
EINECS:200-662-2
UN:1090
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:<–20 °C (closed cup)
REACH:01-2119471330-49
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Assay (GC):≥ 99.9 %
- Solubility in water:Passes test ACS
- Substances reducing KMnO₄ (as O):Passes test ACS
- Acidity:≤ 0.0002 meq/g
- Alkalinity:≤ 0.0002 meq/g
- Colouration:≤ 10 APHA
- Aldehydes (as HCHO):≤ 10 ppm
- Evaporation residue:≤ 5 ppm
- 2-Propanol:≤ 0.05 %
- Methanol:≤ 0.05 %
- Water:≤ 0.2 %
- Absorbance (330 nm):≤ 0.70
- Absorbance (335 nm):≤ 0.22
- Absorbance (340 nm):≤ 0.07
- Absorbance (345 nm):≤ 0.02
- Absorbance (from 350 nm):≤ 0.004
- Transmittance (330 nm):≥ 20 %
- Transmittance (335 nm):≥ 60 %
- Transmittance (340 nm):≥ 85 %
- Transmittance (345 nm):≥ 95 %
- Transmittance (from 350 nm):≥ 99 %
- Fluorescence (as quinine) (365 nm):≤ 1.0 ppb
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
spec_VWRC84700.260 spec_VWRC84700.320
Product specification
Frequently Bought Together