

2-Octanone ≥98.0% (by GC)
: TCI
:111-13-7
:
:C₈H₁₆O
:MFCD00009540
:128.21 g/mol
:14,04711
:173°C
:–16 °C
:0,8171 g/cm³ (20 °C)
:55°C
:Ambient
:203-837-1
:1224
:3,III
:GHS02
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together