2,2,3,3,4,4,5,5,5-Nonafluoro-1-pentanol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 1H,1H-Nonafluoro-1-pentanol, 1H,1H-Perfluoropentan-1-ol
CAS Number:355-28-2
:
Formula:C₅H₃F₉O
MDL Number:MFCD00236714
:250.06 g/mol
Boiling Pt:111°C
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together