Order Entry
Belgium
ContactUsLinkComponent
 
4-(Trifluoromethyl)thiobenzamide ≥98.0% (by HPLC)
 :  TCI
 
undefined
4-(Trifluoromethyl)thiobenzamide ≥98.0% (by HPLC)
 :  TCI

 

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 93.0 %
  • Melting point:
    135.0 to 139.0 deg-C

 

Frequently Bought Together