
Palladium(II) trifluoroacetate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:42196-31-6
:
Formula:C₄F₆O₄Pd
MDL Number:MFCD00013204
:332.45 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together