
Luteolin ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:491-70-3
:
Formula:C₁₅H₁₀O₆
MDL Number:MFCD00017309
:286.24 g/mol
Merck Index:14,05614
Melting Pt:330°C
EINECS:207-741-0
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 94.0 %
Specification Test Results
Frequently Bought Together