Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide concentrate for dilution to 0.5 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Corrosive
Sodium hydroxide concentrate for dilution to 0.5 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 84597.600
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Sodium hydroxide concentrate for dilution to 0.5 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 84597.600
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    210 mL
  • Seq:
    2545

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.4998 0.5002 mol/l

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:>105 °C (1013 hPa)
Density:1.327 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
 :Dose for preparation of 1 litre of solution.
EINECS:215-185-5
UN:1824
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548