p,p,p-Tritolylphosphate ≥98.0% (by GC)
: TCI
:78-32-0
:
:C₂₁H₂₁O₄P
:368.37 g/mol
:430°C
:77 °C
:1,247 g/cm³ (20 °C)
:230 °C
:201-105-6
:3077
:9,III
:GHS07
:GHS09
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:75.0 to 78.0 deg-C
Frequently Bought Together