N-Octyl-β-D-glucopyranoside ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside
CAS Number:29836-26-8
:
Formula:C₁₄H₂₈O₆
MDL Number:MFCD00063288
:292.37 g/mol
Merck Index:14,06767
Melting Pt:110 °C
EINECS:249-887-8
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Specific rotation [a]20/D:-23.0 to -31.0 deg (C=1, methanol)
Specification Test Results
Frequently Bought Together