Order Entry
Belgium
ContactUsLinkComponent
4-Amino-3-hydroxy-1-naphthalenesulphonic acid ≥97.0% (by titrimetric analysis)
4-Amino-3-hydroxy-1-naphthalenesulphonic acid ≥97.0% (by titrimetric analysis)
  TCIAA0367-25G
 :  TCI
 :  
undefined
4-Amino-3-hydroxy-1-naphthalenesulphonic acid ≥97.0% (by titrimetric analysis)
  TCIAA0367-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Brown to Dark purple powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Neutralization titration):
    min. 97.0 % (calcd.on dried substance)
  • Drying loss:
    max. 3.0 %

 

 

 :
 :239.25 g/mol
 :14,00454
 :466 °C (1013 hPa)
 :∼285 °C
 :Ambient
 :204-147-3
 :GHS07
 :116-63-2
 :C₁₀H₉NO₄S
 :MFCD00004019