Order Entry
Belgium
ContactUsLinkComponent
 
Tetrazolium blue chloride ≥95.0% (by titrimetric analysis)
  TCIAD0943-1G
 :  TCI
 :  
undefined
Tetrazolium blue chloride ≥95.0% (by titrimetric analysis)
  TCIAD0943-1G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Amber powder to crystal
  • Purity (HPLC):
    min. 90.0 area%
  • Purity (Nonaqueous Titration):
    min. 95.0 % (calcd.on dried substance)
  • Lambda max.:
    250.0 to 254.0 nm (MeOH)
  • Absorbance (E1%1cm):
    min. 710 (MeOH, 250.0 to 254.0 nm)

 

 

 :
 :14,09244
 :248°C
 :727.65 g/mol
 :Ambient
 :217-488-8
 :1871-22-3
 :C₄₀H₃₂Cl₂N₈O₂
 :MFCD00040933