
Heptafluorobutyric anhydride (HFAA) ≥95.0% (by titrimetric analysis)
Catalog # TCIAH0337-10G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (Anilin Method):min. 95.0 %
Specification Test Results
About this item
:
:410.06 g/mol
Boiling Pt:110°C
Melting Pt:14 °C
Density:1,653 g/cm³ (25 °C)
EINECS:206-410-8
UN:3265
ADR:8,III
:GHS05
CAS Number:336-59-4
Formula:C₈F₁₄O₃
MDL Number:MFCD00000432