DL-α-Phenylalanine ≥98.0% (by HPLC, titration analysis)
TCIAP0136-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in hot Water:almost transparency
:
:165.19 g/mol
:13,07355
:313 °C (1013 hPa)
:173 °C
:205-756-7
:150-30-1
:C₉H₁₁NO₂
:MFCD00064225