Tetrakis(triphenylphosphine)palladium(0) ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:14221-01-3
:
Formula:C₇₂H₆₀P₄Pd
MDL Number:MFCD00010012
:1155.58 g/mol
Melting Pt:103…107 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Freezer
EINECS:238-086-9
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystaline
- Purity (Chelometric Titration):97.0 to 106.0 %
Specification Test Results
Related Information
Frequently Bought Together