

5-Fluorouracile ≥99.0% (par HPLC, analyse par titration)
Supplier: TCI
Certificates
About this item
CAS Number:51-21-8
:
Formula:C₄H₃FN₂O₂
MDL Number:MFCD00006018
:130.08 g/mol
Merck Index:14,04181
Boiling Pt:347 °C (1013 hPa)
Melting Pt:283°C
Storage Temperature:Température ambiante
EINECS:200-085-6
UN:2811
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together