ContactUsLinkComponent
Lecithin (from soy bean)
Lecithin (from soy bean)
Supplier:  TCI
Certificates
Lecithin (from soy bean)
Supplier:  TCI
Certificates
Specification Test Results
  • Appearance:
    Yellow to Amber to Dark purple clear liquid to slightly cloudy liquid
  • Saponification Value:
    180.0 to 195.0
  • Acetone insoluble matter:
    58.0 to 65.0 %
  • Acid Value:
    max. 25.0