
2-Hydroxy-1-naphthoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2283-08-1
:
Formula:C₁₁H₈O₃
MDL Number:MFCD00004009
:188.18 g/mol
EINECS:218-919-2
:GHS07
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together