Order Entry
Belgium
ContactUsLinkComponent
Benzethonium chloride ≥97.0% (by HPLC)
Irritant
Environment
Benzethonium chloride ≥97.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
Irritant
Environment
Benzethonium chloride ≥97.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (TPBNa Method):
    min. 95.0 %
  • Melting point:
    159.0 to 165.0 deg-C
  • Solubility in Water:
    almost transparency

Specification Test Results

Frequently Bought Together