

ω-Bromo-4-methylacetophenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:619-41-0
:
Formula:C₉H₉BrO
MDL Number:MFCD00000203
:213.07 g/mol
Boiling Pt:159°C
Melting Pt:95…97 °C
Storage Temperature:Refrigerator
EINECS:210-595-0
UN:2811
ADR:6.1,III
:GHS05
:GHS06
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:52.0 to 55.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together