
2',4',6'-Trihydroxyacetophenone monohydrate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:480-66-0
:207-556-5
Formula:C₈H₈O₄·1H₂O
MDL Number:MFCD00149091
:186.16 g/mol
Boiling Pt:394 °C (1013 hPa)
Melting Pt:216…218 °C
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:213.0 to 223.0 deg-C
- Water:8.5 to 11.0 %
Specification Test Results
Frequently Bought Together