Order Entry
Belgium
ContactUsLinkComponent
Palladium(II) bromide ≥98.0% (by titrimetric analysis)
Irritant
Palladium(II) bromide ≥98.0% (by titrimetric analysis)
Catalog # TCIAP1937-2G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Palladium(II) bromide ≥98.0% (by titrimetric analysis)
Catalog # TCIAP1937-2G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 g
  • Seq:
    3107

Specifications

Specification Test Results

  • Appearance:
    Light gray to Brown to Black powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :266.23 g/mol
Density:5,173 g/cm³ (16 °C)
EINECS:236-588-2
 :GHS07
CAS Number:13444-94-5
Formula:Br₂Pd
MDL Number:MFCD00011170