3-(Trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)
Catalog # TCIAT2460-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:203.19 g/mol
Boiling Pt:208°C
Density:1,33 g/cm³ (20 °C)
Flash Pt:95°C
UN:2810
ADR:6.1,III
:GHS06
CAS Number:1840-19-3
Formula:C₈H₄F₃NS
MDL Number:MFCD00004806