Order Entry
Belgium
ContactUsLinkComponent
 
 
(2-Chlorophenyl)acetonitrile ≥98.0% (by GC)
 :  TCI
 
undefined
 
(2-Chlorophenyl)acetonitrile ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %
  • Freezing point:
    22.0 to 25.0 deg-C

 

Frequently Bought Together