4-Nitrophenyl isothiocyanate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2131-61-5
:
Formula:C₇H₄N₂O₂S
MDL Number:MFCD00007307
:180.19 g/mol
Boiling Pt:331 °C (1013 hPa)
Melting Pt:108…110 °C
Storage Temperature:Refrigerator
EINECS:218-359-9
UN:2811
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:110.0 to 114.0 deg-C
Specification Test Results
Frequently Bought Together