Methyl hexanoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester
CAS Number:106-70-7
:
Formula:C₇H₁₄O₂
MDL Number:MFCD00009510
Boiling Pt:150°C
Molecular Weight:130.19 g/mol
Melting Pt:-63
Flash Pt:46°C
EINECS:203-425-1
UN:3272
ADR:3 III
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.0 %
- Acid Value:max. 1.0
Specification Test Results
Frequently Bought Together