Order Entry
Belgium
ContactUsLinkComponent
 
 
N-(1-Naphthyl)ethylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen)
 :  TCI
 
undefined
 
N-(1-Naphthyl)ethylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen)
 :  TCI
 

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 % (calcd.on anh.substance)
  • Solubility in Water:
    almost transparency
  • Water:
    max. 7.0 %

 

 

Frequently Bought Together