(±)-2-Tetradecanol ≥95.0% (by GC)
TCIAT0614-10G
: TCI
:
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Environmentally Preferable:
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:33.0 to 35.0 deg-C
- Solubility in Methanol:almost transparency
: 2-Tetradecanol, (±)-2-Hydroxy tetradecane, 2-Tetradecyl alcohol
:
:214.39 g/mol
:170°C
:34 °C
:0,8315 g/cm³ (20 °C)
:225-192-5
:4706-81-4
:C₁₄H₃₀O
:MFCD00004552