



Zinc chloride ≥97.0%, granules, AR® ACS, Macron Fine Chemicals™
:7646-85-7
:
:ZnCl₂
:MFCD00011295
:136.3 g/mol
:13,10185
:732 °C (1013 hPa)
:283 °C
:2.91 g/cm³ (20 °C)
:231-592-0
:2331
:8,III
:GHS05
:GHS07
:GHS09
:GHS08
:Ambient
- For Laboratory,Research,or Manufacturing Use:= 97.0 %
- ACS - Assay (ZnCl₂):≥97.0 %
- Ammonium (NH₄):≤0.005 %
- Calcium (Ca):≤0.06 %
- Insoluble Matter:≤0.005 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Lead (Pb):≤0.005 %
- Magnesium (Mg):≤0.01 %
- Nitrate (NO₃):≤0.003 %
- Oxychloride:Passes Test
- Potassium (K):≤0.02 %
- Sodium (Na):≤0.05 %
- Solubility:Passes Test
- Sulfate (SO₄):≤0.01 %
Frequently Bought Together